missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ENStem-A™ Chemicon™ Neural Freezing Medium (1x)

Catalog No. SCM011
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM011 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM011 Supplier MilliporeSigma™ Supplier No. SCM011
Only null left

qualified for use with ENStem-A Human Neural Progenitor Cells.

ENStem-A™ Neural Freezing Medium is qualified for use with ENStem-A Human Neural Progenitor Cells (Part No. SCC003) cultured in defined serum-free conditions with ENStem-A™ Neural Expansion Medium (Cat. No. SCM004). The optimized formulation allows for consistent cryopreservation and high viability upon thawing and plating.

Specifications

Content And Storage -20°C
For Use With (Application) Stem cell culture
Quantity 50 mL