Learn More
MilliporeSigma™ Empty Monitor Cassette, 25 mm, Type A
Sample collection and particle analysis
Supplier:  MilliporeSigma™ M000025A0
Catalog No. M000025A0
Only null left
Description
Description
Accessory for 3-piece PS monitor, Type A
Order Info
Bulk pack.
Specifications
Specifications
Environmental Test Kits or Supplies
Female Luer slip, requires tubing adapter XX62 000 05
Sample collection, Particle analysis, Environmental analysis
Polystyrene
EMD Millipore Aerosol Analysis Monitor (25 mm)
Empty Monitor Cassette, 25  mm, Type A
Product Suggestions
Product Suggestions