missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ EmbryoMax™ Modified DPBS With BSA and Phenol Red
Specifications
Specifications
| Content And Storage | 2- 8°C up to 2 months |
| For Use With (Application) | Stem cell culture and embryo culture |
| Quantity | 100 mL |