Learn More
MilliporeSigma™ EmbryoMax™ FHM HEPES Buffered Medium (1x)
Optimized and validated for Embryo Culture.
$61.40 - $70.20
Products 4
Catalog Number Mfr. No. Quantity With Additives Without Additives Price Quantity & Availability  
Catalog Number Mfr. No. Quantity With Additives Without Additives Price Quantity & Availability  
MR056F
View Documents Promotion Details
MilliporeSigma™
MR056F
10 mL Phenol Red, Hyaluronidase None
Each for $61.40
Only null left
MR024D
View Documents Promotion Details
MilliporeSigma™
MR024D
50 mL Phenol Red None
Each for $61.40
Only null left
MR122D
View Documents Promotion Details
MilliporeSigma™
MR122D
50 mL None No Phenol Red, No Bovine Serum Albumin (BSA)
Each for $70.20
Only null left
MR025D
View Documents Promotion Details
MilliporeSigma™
MR025D
50 mL None No Phenol Red
Each for $61.40