missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ EmbryoMax™ FHM HEPES Buffered Medium (1x)

Catalog No. MR056F
Change view
Click to view available options
Quantity:
10 mL
50 mL
With Additives:
None
Phenol Red
Phenol Red, Hyaluronidase
Without Additives:
No Phenol Red
No Phenol Red, No Bovine Serum Albumin (BSA)
None
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives
MR056F 10 mL Phenol Red, Hyaluronidase None
MR024D 50 mL Phenol Red None
MR122D 50 mL None No Phenol Red, No Bovine Serum Albumin (BSA)
MR025D 50 mL None No Phenol Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MR056F Supplier MilliporeSigma™ Supplier No. MR056F

Optimized and validated for Embryo Culture.

Specifications

With Additives Phenol Red, Hyaluronidase
Without Additives None
For Use With (Application) Stem Cell Culture
Quantity 10 mL
Content And Storage -20°C