missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ EmbryoMax™ ES Cell 2X Cell Culture Freezing Medium

Catalog No. ES0025F
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES0025F 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES0025F Supplier MilliporeSigma™ Supplier No. ES0025F
Only null left

Formulated with 20% DMSO and Fetal Bovine Serum and available in a 5 x 10mL format.

This unique formulation of cell culture freezing media is for cryopreservation of a broad spectrum of mammalian cells (see viability data). Freezing Media contain 10% of the active cryopreservation ingredient (either DMSO or Glycerol). The remaining 90% is a proprietary mixture of Calf and Fetal Bovine Serum. The reagents used in these products are certified to be virus and mycoplasma free.

Specifications

Content And Storage -80°C up to 2 years
For Use With (Application) Stem cell culture
Quantity 50 mL