missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ EEDα, Mouse monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17663MI
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17663MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17663MI Supplier MilliporeSigma™ Supplier No. 17663

Mouse Monoclonal Antibody

Specifically detects EEDα Clone: in Human, Mouse samples, and it is validated for ChIP, Western Blotting

Specifications

Antigen EEDα
Applications ChIP Assay, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-EED (Mouse monoclonal IgG). One vial containing 50μg of protein G purified antibody in 0.1 M Tris-Glycine (pH 7.4) 150mM NaCl, containing 0.05% azide. May contain 30% glycerol (see certificate of analysis for details). Normal mouse IgG. Two vials containing 25μg of purified mouse IgG in 25μL of storage buffer containing 0.1% sodium azide. ChIP Primers, HoxA2 upstream. One vial containing 75μL of 5μM of each primer specific for the promoter region of human HoxA2. FOR: AGG AAA GAT TTT GGT TGG GAA G; REV: AAA AAG AGG GAA AGG GAC AGA C
Gene Accession No. O75530
Gene Symbols EED; WAIT-1
Host Species Mouse
Immunogen Recombinant fusion protein of full length murine EED tagged with MBP.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003788
Test Specificity Recognizes EED, MW: ∽50kDa.
Includes This ChIPAb+ EED -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human, Mouse
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only