missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ RESGRO™ Culture Medium

a specialty media that can be utilized to complement traditional murine Embryonic Stem (ES) cell culture media containing ESGRO murine LIF medium supplement.

Specifications

For Use With (Application) Stem cell culture
Content And Storage −20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SCM001
View Documents Promotion Details
MilliporeSigma™
SCM001
250 mL N/A
Only null left
 
SCM002
View Documents Promotion Details
MilliporeSigma™
SCM002
500 mL N/A
Only null left
 
Description

Description

RESGRO Culture Medium is a specialty media that can be utilized to complement traditional murine Embryonic Stem (ES) cell culture media containing ESGRO™ murine LIF medium supplement. In contrast to routine ES cell culture using ESGRO™ supplemented medium, RESGRO Culture Medium is recommended for a number of specialized applications.
Specifications

Specifications

Stem cell culture
−20°C
Videos
SDS
Documents

Documents