missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ ReNcell Neural Stem Cell Freezing Medium
Description
ReNcell Neural Stem Cell Freezing Medium is qualified for use with ReNcell immortalized human neural progenitor cell lines, CX (CHEMICON Catalog No. SCC007) and VM (CHEMICON Catalog No. SCC008) cultured in serum-free conditions with ReNcell NSC Maintenance Medium (CHEMICON Catalog No. SCM005). The optimized formulation allows for consistent cryopreservation and high viability upon thawing and plating.
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Application) | Stem cell culture |
| Quantity | 50 mL |