missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Recombinant Rat Leukemia Inhibitory Factor

Catalog No. LIF3010
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LIF3010 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LIF3010 Supplier MilliporeSigma™ Supplier No. LIF3010

Recombinant rat Leukemia Inhibitory Factor (LIF) protein is biologically active and suitable for cell culture applications.

Leukaemia inhibitory factor is a cytokine that induces macrophage differentiation. Neurotransmitters and neuropeptides, well known for their role in the communication between neurons, are also capable of activating monocytes and macrophages and inducing chemotaxis in immune cells. LIF signals through different receptors and transcription factors. LIF in conjunction with BMP2 acts in synergy on primary fetal neural progenitor cells to induce astrocytes.

Specifications

Content And Storage -20°C
For Use With (Application) Cell culture
Quantity 100 μg
Source E. coli