missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Recombinant Mouse Leukemia Inhibitory Factor Protein

Catalog No. LIF2010
Change view
Click to view available options
Quantity:
10 μg
5 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LIF2010 10 μg
LIF2005 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LIF2010 Supplier MilliporeSigma™ Supplier No. LIF2010

biologically active and suitable for cell culture applications.

Leukaemia inhibitory factor is a cytokine that induces macrophage differentiation. Neurotransmitters and neuropeptides, well known for their role in the communication between neurons, are also capable of activating monocytes and macrophages and inducing chemotaxis in immune cells. LIF signals through different receptors and transcription factors. LIF in conjunction with BMP2 acts in synergy on primary fetal neural progenitor cells to induce astrocytes.

Specifications

Content And Storage 2- 8°C for 6 - 12 months
For Use With (Application) Cell culture and stem cell culture
Quantity 10 μg
Source E. coli