missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Rat Neural Stem Cell Expansion Medium

Catalog No. SCM009
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM009 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM009 Supplier MilliporeSigma™ Supplier No. SCM009

provided as a two-component system that is convenient and easy to use.

The Rat Neural Stem Cell Expansion Medium is provided as a two-component system that is convenient and easy to use. This product includes the Neural Stem Cell Basal Medium, a defined serum-free medium that has been optimized for the growth and in vitro differentiation of neural stem cells derived from rodents. A solution of basic FGF-2 growth factor is also provided separately, which when added to the Neural Stem Cell Basal Medium allows for the growth and proliferation of rat neural stem cells. Withdrawal of bFGF-2 from the Neural Stem Cell Basal Medium will result in the spontaneous differentiation of rat neural stem cells.

Specifications

Content And Storage Neural Stem Cell Basal Medium (Mfr. No. SCM003), 500mL; bFGF, 10μg. Store at -20°C
For Use With (Application) Stem cell culture
Quantity 1 Kit