missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ MTT Cell Growth Assay Kit
For proliferation or complement-mediated cytotoxicity assays
Specifications
| Content And Storage | 2°C to 8°C |
|---|---|
| Description | MTT Cell Growth Assay Kit |
| Detection Method | Colorimetric |
| For Use With (Application) | For proliferation or complement-mediated cytotoxicity assays |
| Includes | Solution B (PBS), Reagent A (MTT) |
Specifications
| 2°C to 8°C | |
| Colorimetric | |
| Solution B (PBS), Reagent A (MTT) |
| MTT Cell Growth Assay Kit | |
| For proliferation or complement-mediated cytotoxicity assays |