missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Mesenchymal Stem Cell Freezing Medium (1x)

Catalog No. SCM016
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM016 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM016 Supplier MilliporeSigma™ Supplier No. SCM016

optimized and validated for Stem cell culture. This medium is available in a 50ml format.

Specifications

Content And Storage -20°C
Serum Level Standard Serum
For Use With (Application) Cryopreservation of human and rodent mesenchymal stem cell lines
Quantity 50 mL