missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ In Vitro Vascular Permeability Assay
For evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability
$950.00 - $1,040.00
Specifications
| Content And Storage | Store kit materials at 2-8°C; use within 4 months from date of receipt |
|---|---|
| Description | In Vitro Vascular Permeability Assay |
| Detection Method | Fluorescence |
| For Use With (Application) | Evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability |
Specifications
| Store kit materials at 2-8°C; use within 4 months from date of receipt | |
| In Vitro Vascular Permeability Assay | |
| Fluorescence | |
| Evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability |