missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ In Vitro Vascular Permeability Assay

Catalog No. ECM644
Change view
Click to view available options
Quantity:
24 Assays
96 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM644 24 Assays
ECM642 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ECM644 Supplier MilliporeSigma™ Supplier No. ECM644

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability

  • Fluorometric detection format.
  • Supports assessment of compounds that disrupt or protect an endothelial monolayer.

Specifications

Content And Storage Store kit materials at 2-8°C; use within 4 months from date of receipt
Description In Vitro Vascular Permeability Assay
Detection Method Fluorescence
For Use With (Application) Evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability
Includes Black plate, 24-well opaque/permeability insert assembly collagen-coated, receiver tray, FITC-dextran solution, cell stain, forceps
Quantity 24 Assays
Druggable Target Cell-based Assay