Learn More
MilliporeSigma™ Chemicon Human OPC Expansion Culture Media Kit
a defined, serum-free medium that has been optimized for the culture and expansion of human ES derived oligodendrocyte progenitor cells.
Supplier:  MilliporeSigma™ SCM107
Catalog No. SCM107
Only null left
Explore available promotions
Description
Description
Human OPC Expansion Media is a defined, serum-free medium that has been optimized for the culture and expansion of human ES derived oligodendrocyte progenitor cells (Catalog No. SCR600). Cells double every 48 hours when cultured in Human OPC Expansion Complete Media. It is not known whether the media can support the culture of primary oligodendrocyte progenitor cells isolated from brain tissues.
Specifications
Specifications
-20°C
Stem Cell Culture
100 mL
Product Suggestions
Product Suggestions