missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ ESGRO™ Leukemia Inhibitory Factor (LIF), 10 million units/1mL
Description
Leukaemia inhibitory factor is a cytokine that induces macrophage differentiation. Neurotransmitters and neuropeptides, well known for their role in the communication between neurons, are also capable of activating monocytes and macrophages and inducing chemotaxis in immune cells. LIF signals through different receptors and transcription factors. LIF in conjunction with BMP2 acts in synergy on primary fetal neural progenitor cells to induce astrocytes.
Specifications
Specifications
| Form | Purified |
| For Use With (Application) | Stem cell culture |
| Quantity | 1 mL |