missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon Endotoxin-Free Dulbecco's PBS (1X) (w/o Ca++ and Mg++)

Catalog No. TMS012A
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TMS012A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TMS012A Supplier MilliporeSigma™ Supplier No. TMS012A

For Cell Culture

Specifications

Content And Storage Room temperature
For Use With (Application) Cell culture
Quantity 1 L