missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Choline Acetyltransferase Blocking Peptide

Catalog No. AG220MI
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG220MI 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG220MI Supplier MilliporeSigma™ Supplier No. AG220

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Blocking AB143, AB144P Antibodies

Liquid in 50mM KPO4 buffer pH 7.5 containing 10% glycerol and 0.05M NaCL. Contains no preservative.

Recombinant rat ChAT

Recommended Storage

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Specifications

Chemical Name or Material Choline Acetyltransferase Blocking Peptide
Concentration 0.25 mg/mL
For Use With (Application) Cell Function Assay
Quantity 10 μg