missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ AOF ITS Supplement (100x)

Catalog No. SCM055
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM055 100 mL
SCM054 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SCM055 Supplier MilliporeSigma™ Supplier No. SCM055

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

an animal-origin-free product that contains human recombinant insulin, transferrin along with sodium selenite and is used for the serum-free culture of numerous cell types.

Specifications

Content And Storage 2-8°C
For Use With (Application) Stem cell culture, cell culture, cell stimulation, cell growth
Quantity 100 mL