missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Accutase™ Cell Detachment Solution

Catalog No. SCR005
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR005 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR005 Supplier MilliporeSigma™ Supplier No. SCR005
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.

Useful for the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware, including SmartPlastic™. ACCUTASE™ does not contain mammalian or bacterial derived products.

Specifications

Content And Storage -20
For Use With (Application) Creating single cell suspensions from clumped cell cultures 
Quantity 100 mL