missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Accumax™ Cell Detachment Solution

Catalog No. SCR006
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR006 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR006 Supplier MilliporeSigma™ Supplier No. SCR006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A cell detachment solution of proteolytic, collagenolytic and DNAse enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.

Useful on a variety of cells lines however not all cells or cell types are affected by ACCUMAX™, also passage number and confluency can influence how well the reagent does because the amounts and makeup of the collagen extracellular matrix will vary with age and culture state. Because ACCUMAX™ is a more gentle, less univerally proteolytic cleavage mixture reagent amounts and incubation times need to be optimized for each cell system.

Specifications

Content And Storage -20
For Use With (Application) Creating single cell suspensions from clumped cell cultures 
Quantity 100 mL