missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Cell Culture Freezing Media with DMSO

Catalog No. p-5235433
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S002D 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S002D Supplier MilliporeSigma™ Supplier No. S002D
Only null left

Optimized and validated for cell culture and Stem cell culture.

This unique formulation of cell culture freezing media is for cryopreservation of a broad spectrum of mammalian cells (see viability data). The reagents used in these products are certified to be virus and mycoplasma free.

Specifications

Content And Storage -20°C
Form Liquid
Formulation 10% DMSO, calf and fetal bovine serum
For Use With (Application) Stem cell culture and cell culture
Quantity 50 mL