missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Casein, Bovine Milk, Carbohydrate and Fatty Acid Free, Protein

Catalog No. 218682500GM
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
218682500GM 500 g
218682100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 218682500GM Supplier MilliporeSigma™ Supplier No. 218682500GM
Only null left
High molecular weight phosphoprotein. Useful as a substrate for bromelain, chymotrypsin, pepsin, trypsin, and other proteases. Supplied as Hammarsten grade material.

Specifications

Preparation Method Dilute Alkaline Solutions (10mg/mL)
Purification Method Assay
Quantity 500 g
Immunogen Casein
Storage Requirements Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C. After one week, it is recommended that fresh solution be prepared.
Regulatory Status RUO
Without Additives Carbohydrate and Fatty Acid Free
Species Bovine
Recombinant Native
Form Solid
Purity or Quality Grade ≥95%
Show More Show Less