Learn More
MilliporeSigma™ Casein, Bovine Milk, Carbohydrate and Fatty Acid Free, Protein
Supplier:  MilliporeSigma™ 218682100GM
 View more versions of this product
Catalog No. 21-868-2100GM
Only null left
Explore available promotions
Description
Description
High molecular weight phosphoprotein. Useful as a substrate for bromelain, chymotrypsin, pepsin, trypsin, and other proteases. Supplied as Hammarsten grade material.
Specifications
Specifications
Dilute Alkaline Solutions (10mg/mL)
100 g
Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C. After one week, it is recommended that fresh solution be prepared.
Carbohydrate and Fatty Acid Free
Native
≥95%
Assay
Casein
RUO
Bovine
Solid
Product Suggestions
Product Suggestions