missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Urease From Canavalia ensiformis

Catalog No. 66613310KU
Change view
Click to view available options
Quantity:
10 KU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66613310KU 10 KU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66613310KU Supplier MilliporeSigma™ Supplier No. 66613310KU

Native urease from Canavalia ensiformis. Catalyzes hydrolysis of urea to carbon dioxide and ammonia. Useful in the determination of urea in body fluids. Note: 1 KU = 1000 units.

Native urease from

Specifications

Content And Storage -20°C
For Use With (Application) Determination of urea in body fluids
Quantity 10 KU