missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Tryptase From Human Lung

Catalog No. 65036625UG
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65036625UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65036625UG Supplier MilliporeSigma™ Supplier No. 65036625UG

Native tryptase from human lung.

Tryptase is a serine protease stored in mast cell granules. A tetrameric enzyme composed of four non-covalently bound subunits. Each subunit is reported to contain one active site. Reported to upregulate neoangiogenesis in carcinogenesis of the uterine cortex. Tryptase is also an important mediator of allergic reactions that activates G-protein-coupled receptors through proteolysis, thus amplifying histamine release.

Specifications

Content And Storage -20°C
For Use With (Application) Mediator of allergic reactions that activates G-protein-coupled receptors through proteolysis, thus amplifying histamine release
Quantity 25 μg
Source Human lung