missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Trypsin Porcine MS Grade

Catalog No. 6502791SET
Change view
Click to view available options
Quantity:
5 x 20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6502791SET 5 x 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6502791SET Supplier MilliporeSigma™ Supplier No. 6502791SET

A chemically modified trypsin that is resistant to autolysis and degradation

Trypsin specifically hydrolyzes amides, peptides and proteins at the C-terminus side of arginine and lysine residues. This mass spectrometry grade trypsin is methylated, TPCK treated, and affinity purified. It is specifically designed for digestion of proteins for sequencing and peptide fragment analysis by mass spectrometry. A trypsin to protein ratio of 1:100 to 1:20 is recommended for mass spectrometry protocols. Note: 1 SET = 5 X 20μg of modified trypsin, and 1mL of resuspension buffer.

Specifications

Content And Storage -20°C
For Use With (Application) Digestion of proteins for sequencing and peptide fragment analysis by mass spectrometry
Quantity 5 x 20 μg
Source Porcine