missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Trypsin Iodination Grade From Human Pancreas

Catalog No. 65027550UG
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65027550UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65027550UG Supplier MilliporeSigma™ Supplier No. 65027550UG

Native trypsin from human pancreas.

Serine protease that specifically hydrolyzes amides, peptides, and proteins at the C-terminus side of arginine and lysine residues.

Specifications

Content And Storage -20°C
Formulation Salt-free
For Use With (Application) Hydrolyzes amides, peptides and proteins at C-terminal side of arginine and lysine residues
Quantity 50 μg
Source Human pancreas