missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Trypsin From Bovine Pancreas

Catalog No. 650225MU
Change view
Click to view available options
Quantity:
25 mU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
650225MU 25 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 650225MU Supplier MilliporeSigma™ Supplier No. 650225MU

Native trypsin from bovine pancreas.

Serine protease that specifically hydrolyzes amides, peptides and proteins at the C-terminal side of arginine and lysine residues. Inhibitors include aprotinin, DFP, and soybean trypsin inhibitor. Note: 1 MU = 1,000,000 units.

Specifications

Content And Storage -20°C
Formulation Salt-free
For Use With (Application) Hydrolyzes amides, peptides and proteins at C-terminal side of arginine and lysine residues
pH Optimum 8.5 to 8.8
Quantity 25 mU
Source Bovine pancreas