missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Human Thyroxine-Binding Globulin
Specifications
Specifications
| Content And Storage | -20°C |
| Formulation | In 140mM NaCl, 10mM Tris, 0.1% NaN3, pH 8.0. Sterile-filtered |
| Quantity | 250 μg |
| Source | Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV |