missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Human Plasma beta2-Glycoprotein 1

Catalog No. 362225100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
362225100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 362225100UG Supplier MilliporeSigma™ Supplier No. 362225100UG

Native β2-glycoprotein 1 from human plasma.

A carbohydrate-rich plasma protein that is a constituent of several lipoprotein particles. Possesses sialylated and galactosylated bi- and triantennary oligosaccharides.

Specifications

Content And Storage 2-8°C
Formulation Lyophilized from 200mM glycine, 150mM NaCl, pH 7.5
Quantity 100 μg