missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Human Plasma alpha2-HS-Glycoprotein

Catalog No. 3621991MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3621991MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3621991MG Supplier MilliporeSigma™ Supplier No. 3621991MG

Native α2-HS-glycoprotein from human plasma.

Plasma protein that is also found in the extracellular matrix. Synthesized in liver and enriched in bone. Its concentration dynamically changes under various physiological conditions. Functions as an opsonin for cationic-deactivating molecules in macrophages.

Specifications

Content And Storage -20°C
Formulation Lyophilized from 200mM NaCl, 20 mM Tris-HCl, pH 8.0
Quantity 1 mg