missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Freund's Incomplete Adjuvant, Modified

Catalog No. 34429110ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34429110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34429110ML Supplier MilliporeSigma™ Supplier No. 34429110ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Yellow oily heat sterilized liquid

  • Composition: 10mL of Freund's Incomplete Adjuvant homogenized in the following amounts by weight: 85% Drakeol 5NF, 15% Arlacel A (mannide monooleate emulsifier)
  • Heat sterilized
  • Contains zero colony forming units/mL

Specifications

Product Type Freund's Incomplete Adjuvant, Modified
Quantity 10 mL
Content And Storage 15°C to 30°C