missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ CELLULYSIN™ Cellulase
Description
Hydrolyzes β1,4-glucan linkages in cellulose. Has been used to isolate intact cells and protoplasts from plant tissues.
Specifications
Specifications
| Content And Storage | 2-8°C |
| For Use With (Application) | Isolate intact cells and protoplasts from plant tissues |
| Quantity | 50 KU |