missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Avidin, Egg White
Glycoprotein containing four identical subunits
$138.68 - $138.68
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-972-510MG
|
MilliporeSigma™
18972510MG |
10 mg |
Each for $138.68
|
|
|||||
Description
- Form: White lyophilized solid
- Molar Mass: 66,000
- Biological Activity: ≥10 units/mg protein
- Soluble in dH2O and dilute buffer
- Storage: Store at -20°C
Specifications
| -20°C |