Learn More
MilliporeSigma™ BrdU Cell Proliferation Assay Kits
BrdU Cell Proliferation Assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells. It is sensitive, rapid, and easy to perform.
Supplier: MilliporeSigma™ QIA581000TEST
Description
BrdU Cell Proliferation Assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells. It is sensitive, rapid, and easy to perform.
Kit Components:
- BrdU Label 58
- Fixative/Denaturing Solution 58
- Anti-BrdU Antibody 58
- Antibody Diluent 58
- TMB (Soluble)
- ELISA Conjugate Diluent(-20C, 4%BSA)
- ELISA Stop Solution
- ELISA 20X Plate Wash Concentrate
- Peroxidase Goat anti-Mouse IgG
Chemical Identifiers
| 96-well plate | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| Non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells |
Specifications
| 96-well plate | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| 1000 Tests | |
| Non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells |
For Research Use Only