missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Immobilon™ Block Noise Canceling Reagents
Minimize background in Western blots, increasing sensitivity for superior detection of even low-abundance proteins
$226.00 - $254.18
Specifications
| Quantity | 500 mL |
|---|---|
| Sufficient For | Nineteen 8 x 7cm miniblots |
| Content And Storage | 20°C to 25°C |
| Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Application) | Product Type | Price | Quantity | ||||
|
WBAVDCH01
|
MilliporeSigma™
WBAVDCH01 |
Mass Spectrometry | Block-CH Reagent |
Each for $242.00
|
|
||||
|
WBAVDFL01
|
MilliporeSigma™
WBAVDFL01 |
Fluorescence Assay | Block-FL Reagent |
Each for $226.00
|
|
||||
|
WBAVDP001
|
MilliporeSigma™
WBAVDP001 |
Mass Spectrometry | Block-PO Reagent |
Each for $254.18
|
|
||||
Description
- Ready-to-use
- Protein-free, reducing nonspecific binding, especially when using phosphoantibodies or avidin-biotin systems
- Specially engineered for the EMD Millipore SNAP i.d.™ detection system, but may be used in traditional Western blotting protocols
- Suitable for the dilution of primary and secondary antibodies
- Sufficient for 19 miniblots (each 8 × 7cm blot requires at least 26mL for SNAP i.d.)
- Compatible with streptavidin antibodies
- Compatible with PVDF (Immobilon™ P, Immobilon FL) and nitrocellulose
- Block-CH reagent is compatible with Immobilon HRP substrate
- Choose from three Block reagents
- Block-CH reagent for HRP chemiluminescence detection
- Block-FL reagent for fluorescence detection
- Block-PO reagent for phosphoprotein detection via chemiluminescence or fluorescence
Specifications
| 500 mL | |
| Nineteen 8 x 7cm miniblots | |
| 20°C to 25°C |