missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Immobilon™ Block Noise Canceling Reagents

Catalog No. WBAVDCH01
Change view
Click to view available options
For Use With (Application):
Fluorescence Assay
Mass Spectrometry
Product Type:
Block-CH Reagent
Block-FL Reagent
Block-PO Reagent
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Product Type
WBAVDCH01 Mass Spectrometry Block-CH Reagent
WBAVDFL01 Fluorescence Assay Block-FL Reagent
WBAVDP001 Mass Spectrometry Block-PO Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. WBAVDCH01 Supplier MilliporeSigma™ Supplier No. WBAVDCH01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Minimize background in Western blots, increasing sensitivity for superior detection of even low-abundance proteins

 

  • Ready-to-use
  • Protein-free, reducing nonspecific binding, especially when using phosphoantibodies or avidin-biotin systems
  • Specially engineered for the EMD Millipore SNAP i.d.™ detection system, but may be used in traditional Western blotting protocols
  • Suitable for the dilution of primary and secondary antibodies
  • Sufficient for 19 miniblots (each 8 × 7cm blot requires at least 26mL for SNAP i.d.)
  • Compatible with streptavidin antibodies
  • Compatible with PVDF (Immobilon™ P, Immobilon FL) and nitrocellulose

 

 

  • Block-CH reagent is compatible with Immobilon HRP substrate
  • Choose from three Block reagents
  • Block-CH reagent for HRP chemiluminescence detection
  • Block-FL reagent for fluorescence detection
  • Block-PO reagent for phosphoprotein detection via chemiluminescence or fluorescence

 

Specifications

Product Type Block-CH Reagent
For Use With (Application) Mass Spectrometry
Quantity 500 mL
Sufficient For Nineteen 8 x 7cm miniblots
Content And Storage 20°C to 25°C