missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Animal Free Collagenase/Dispase Blend II

Catalog No. SCR140
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR140 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR140 Supplier MilliporeSigma™ Supplier No. SCR140

missing translation for 'validMainframeMsg'

a mixture of animal free collagenase, type B, and neutral protease/dispase, and is inteneded to be used when a more potent enzymatic dissociatation solution than collagnaease is desired.

Specifications

Content And Storage 2-8°C
For Use With (Application) Cell cuture and cell harvesting
Quantity 50 mg
Source Clostridium histolyticum and Bacillus polymyxa