missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Animal Free Collagenase/Dispase Blend I
Specifications
Specifications
| Content And Storage | 2-8°C |
| For Use With (Application) | Cell cuture and cell harvesting |
| Quantity | 50 mg |
| Source | Clostridium histolyticum and Bacillus polymyxa |