missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Angiostatin K1-3, Human, Recombinant Protein, E. coli

Catalog No. 176708250ug
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
176708250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 176708250UG Supplier MilliporeSigma™ Supplier No. 176708250UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant, human angiostatin expressed in E. coli as a single, nonglycosylated polypeptide chain containing 259 amino acids. Represents a proteolytic fragment containing the first three kringle structures of plasminogen. Acts as a specific inhibitor of endothelial cell growth and angiogenesis. Displays increased inhibitory activity (ED50=70nM) relative to kringles 1-4 (ED50=135nm).

Specifications

Formulation Lyophilized from 20mM Sodium Acetate, 4% Mannitol, pH 5.5.
Preparation Method H2O or Aqueous Buffer Containing 0.1% BSA
Purification Method HPLC and SDS-PAGE
Quantity 250 μg
Source E. coli
Immunogen Angiostatin K1-3
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less