missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ Androgen Receptor- ChIP Validated, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710489MI
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
1710489MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1710489MI Supplier MilliporeSigma™ Supplier No. 1710489

Rabbit Polyclonal Antibody

Specifically detects Androgen Receptor- ChIP Validated Clone: in Amphibian, Bovine, Chicken, Human, Mouse, Rat samples, and it is validated for Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Western Blotting

The androgen receptor (AR) is a type of nuclear receptor which is activated by binding of either of the androgenic hormones testosterone or dihydrotestosterone. The main function of the androgen receptor is as a DNA binding transcription factor which regulates gene expression. However the androgen receptor also has additional functions independent of DNA binding. The androgen receptor is most closely related to the progesterone receptor, and progestins in higher dosages can block the androgen receptor.

Specifications

Antigen Androgen Receptor- ChIP Validated
Applications ChIP Assay, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Androgen Receptor (Rabbit Polyclonal). One vial containing 125μL (0.2μg/μL) purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl, and 0.05% sodium azide, before the addition of 30% glycerol. Normal Rabbit IgG. One vial containing 125μg (1.0μg/μL) Rabbit IgG in 125μL storage buffer containing 0.05% sodium azide. ChIP Primers, PSA. One vial containing 75μL of 5μM of each primer specific for human prostate-specific antigen enhancer region. (chr19:51354179+51354263, hg19 build) FOR: 5’ GCC TGG ATC TGA GAG AGA TAT CAT C 3’; REV: 5’ ACA CCT TTT TTT TTC TGG ATT GTT G 3’
Gene Accession No. P10275
Gene Symbols AR, DHTR, SMAX1, HUMARA, KD, SBMA, TFM, OTTHUMP00000061928, AIS, NR3C4
Host Species Rabbit
Immunogen Peptide corresponding to amino acids 1-21 of the Human Androgen Receptor (MEVQLGLGRVYPRPPSKTYRG)
Purification Method Affinity Purified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_000035
Test Specificity Recognizes Androgen Receptor. No cross-reactivity with estrogen, progesterone or glucocorticoid receptors.
Includes ChIPAb+ Androgen Receptor set includes the Androgen Receptor antibody, a Normal Rabbit IgG, and control primers, which amplify a 85bp region of the human prostate specific antigen (PSA) enhancer region.
Target Species Amphibian, Bovine, Chicken, Human, Mouse, Rat
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Show More Show Less

For Research Use Only