missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Alcian-Blue Staining Solution

Catalog No. TMS010C
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TMS010C 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TMS010C Supplier MilliporeSigma™ Supplier No. TMS010C

often used to identify chondrocytes from differentiated cultures of both human and rodent mesenchymal stem cells (MSCs).

Specifications

For Use With (Application) Stem cell culture, cell based assays, cell differentiation
pH 2.5
Quantity 100 mL