missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Activin A, Human, Recombinant Protein, CHO Cells
Supplier: MilliporeSigma™ 11470010UG
Description
Recombinant, human activin A (disulfide-linked homodimer of two 116-amino acid residue βA subunits) expressed in CHO cells. Mature βA-monomer lacks a potential N-linked glycosylation site and has a calculated molecular weight of 13kDa. A member of the TGF-β superfamily that is involved in the negative regulation of B lineage lymphocytes. Shown to play an important role in the regulation of hormonogenesis, cellular homeostasis, and differentiation programs. Reported to inhibit endogenous production of IL-6.Specifications
| Lyophilized from sterile filtered 35% acetonitrile, 0.1% TFA, 50μg BSA/μg Activin A. | |
| SDS-PAGE | |
| Activin A | |
| RUO | |
| Human | |
| Lyophilized |
| Reconstitute to a concentration ≥10μg/mL with sterile PBS containing ≥0.1% HSA or BSA. | |
| 10 μg | |
| Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 3 months at −70°C. Avoid freeze/thaw cycles of solutions. | |
| CHO Cells | |
| Recombinant | |
| ≥95% |