missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ Acetyl-Histone H4, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 17630MI
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17630MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17630MI Supplier MilliporeSigma™ Supplier No. 17630

Rabbit Polyclonal Antibody

Specifically detects Acetyl-Histone H4 Clone: in Eukaryote, Human samples, and it is validated for Chromatin Immunoprecipitation, Functional Assay, Multiplexing, Western Blotting

Specifications

Antigen Acetyl-Histone H4
Applications Multiplex, Functional Assay, Immunoassay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Acetyl-Histone H4 (rabbit polyclonal IgG). One vial containing 100μL of antiserum containing 0.05% sodium azide. Normal Rabbit Serum. One vial containing 100μL antiserum containing 0.05% sodium azide. Control Primers. One vial containing 75μL of 5μM of each primer specific for for human GAPDH.
Gene Accession No. P62805
Host Species Rabbit
Immunogen The acetyl-histone H4 antiserum is made against a peptide corresponding to amino acids 2-19 of Tetrahymena histone H4.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_778224
Test Specificity Recognizes acetylated histone H4 of approximately 10kDa. Cross-reacts with acetylated histone H2B from Tetrahymena and weakly crossreacts with acetylated histone H2B from HeLa cells. May cross-react with other acetylated proteins.
Includes This ChIPAb+ Acetyl-Histone H4 -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Eukaryote, Human
Content And Storage −20°C from date of receipt
Form Serum
Show More Show Less

For Research Use Only