missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ Acetyl-Histone H4 (Lys8), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710099MI
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
1710099MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1710099MI Supplier MilliporeSigma™ Supplier No. 1710099

Rabbit Polyclonal Antibody

Specifically detects Acetyl-Histone H4 (Lys8) Clone: in Human, S. cerevisiae, Yeast samples, and it is validated for Chromatin Immunoprecipitation, Immunoprecipitation, Western Blotting

Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the 'beads on a string' structure. The N-terminal tail of histone H4 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specifications

Antigen Acetyl-Histone H4 (Lys8)
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Anti-Acetyl-Histone H4 (Lys8) (rabbit polyclonal). One vial containing 50μL of antiserum containing 0.05% sodium azide. Normal Rabbit Serum. One vial containing 100μL of antiserum containing 0.05% sodium azide. ChIP Primers, HSPCA. One vial containing 75μL of 5μM of each primer specific for human HSPCA. FOR: GCA ACA GCT ACC ACA GGA CCA; REV: GAG CGT GTG AAA TCA ACA TAA AGC
Gene Accession No. P62805
Gene Symbols HIST1H4A, H4/J, HIST1H4F, HIST1H4L, H4FN, H4FA, H4FH, HIST1H4H, H4F2, H4FM, H4/A, H4FK, H4FG, HIST2H4, H4FB, H4/K, HIST1H4I, H4/N, HIST1H4B, H4FE, H4/M, H4/a, HIST1H4E, HIST4H4, HIST2H4A, H4FC, H4FI, H4/H, HIST1H4C, H4/G, HIST1H4K, H4FJ, H4FD, H4/I, H4/B, H4/D, H4/C, HIST1H4J, HIST1H4D, H4/E
Host Species Rabbit
Immunogen Synthetic peptide corresponding to yeast Histone H4 acetylated at lysine 8.
Purification Method Unpurified
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NM_175054
Test Specificity Recognizes acetyl-Histone H4 (Lys8).
Includes The ChIPAb+ Acetyl-Histone H4 (Lys8) set includes the Acetyl-Histone H4 (Lys8) antibody, a negative control rabbit serum and qPCR primers which amplify a 134bp region of human HSPCA.
Target Species Human, S. cerevisiae, Yeast
Content And Storage -20°C for up to 12 months from date of receipt; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Serum
Show More Show Less

For Research Use Only