missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Upstate™ Acetyl-Histone H4 (Lys5)α, Rabbit Monoclonal, ChIP Validated Antibody and Primer Set

Catalog No. 1710045MI
Change view
Click to view available options
Quantity:
25 Assays
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
1710045MI 25 Assays Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1710045MI Supplier MilliporeSigma™ Supplier No. 1710045

Rabbit Monoclonal Antibody

Specifically detects Acetyl-Histone H4 (Lys5)α Clone: in Human samples, and it is validated for Chromatin Immunoprecipitation, Immunoprecipitation, Western Blotting

Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the 'beads on a string' structure. Acetylation of histone H4 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs).

Specifications

Antigen Acetyl-Histone H4 (Lys5)α
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Anti-Acetyl-Histone H4 (Lys5) (rabbit monoclonal IgG). One vial containing 50μL of rabbit monoclonal IgG cell culture supernatant in 0.1% sodium azide. Negative Control Supernatant (rabbit). One vial containing 50μL of cultured supernatant in 0.05% sodium azide. Control Primers. One vial containing 75μL of 5μM each primer specific for the human GAPDH promoter. FOR: TAC TAG CGG TTT TAC GGG CG; REV: TCG AAC AGG AGG AGC AGA GAG CGA
Gene Accession No. P62805
Gene Symbols HIST1H4A, H4/J, HIST1H4F, HIST1H4L, H4FN, H4FA, H4FH, HIST1H4H, H4F2, H4FM, H4/A, H4FK, H4FG, HIST2H4, H4FB, H4/K, HIST1H4I, H4/N, HIST1H4B, H4FE, H4/M, H4/a, HIST1H4E, HIST4H4, HIST2H4A, H4FC, H4FI, H4/H, HIST1H4C, H4/G, HIST1H4K, H4FJ, H4FD, H4/I, H4/B, H4/D, H4/C, HIST1H4J, HIST1H4D, H4/E
Host Species Rabbit
Immunogen Peptide corresponding to Histone H4 containing the sequence [GRG-AcK-GGK] on which Lys5 is acetylated.
Quantity 25 Assays
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001029249
Test Specificity Recognizes Histone H4 acetylated on Lys5.
Includes This ChIPAb+ Acetyl-Histone H4 (Lys5) -ChIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.
Target Species Human
Content And Storage -20°C for one year from date of shipment; Upon first thaw and prior to removing the cap centrifuge the vial and gently mix the solution; Aliquot into microcentrifuge tubes and store at −20°C; Avoid repeated freeze/thaw cycles
Form Culture Supernatant
Isotype IgG
Show More Show Less

For Research Use Only