Learn More
MilliporeSigma™ Stainless Steel Plug, 1/4 in. NPT
Accessories for filter holders for sample preparation.
Supplier:  MilliporeSigma™ YY1301009
Catalog No. YY1301009
Only null left
Explore available promotions
Description
Description
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel. Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Specifications
Specifications
Not Autoclavable
Stainless Steel
Plug
Dispensing Pressure Vessel
Non-sterile
2.5 mm
Product Suggestions
Product Suggestions